missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophene-2-carboxaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 11470893
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18791-75-8
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005431
InChI Key PDONIKHDXYHTLS-UHFFFAOYSA-N
Synonym 4-bromothiophene-2-carboxaldehyde, 4-bromo-2-thiophenecarboxaldehyde, 4-bromo-2-formylthiophene, 4-bromo-2-thiophenecarbaldehyde, 4-bromo-thiophene-2-carbaldehyde, 4-bromo-2-thiophene carboxaldehyde, 4-bromothiophen-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4-bromo, 4-bromothiophene-2-aldehyde, 3-bromothiophene-5-carboxaldehyde
PubChem CID 87792
IUPAC Name 4-bromothiophene-2-carbaldehyde
SMILES BrC1=CSC(C=O)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 114°C to 115°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 191.05
Percent Purity 96%
Chemical Name or Material 4-Bromothiophene-2-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'