Learn More
4-(Aminomethyl)pyridine, 97+%, Thermo Scientific Chemicals Product Code.: 11382476
4-(Aminomethyl)pyridine, 97+%, Thermo Scientific Chemicals
Product Code. 11382476
25 g
Click to view available options
Quantity:
25 g
100 g
Product Code. 11382476
Brand: Thermo Scientific Alfa Aesar L06345.14
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3731-53-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006449
InChI Key TXQWFIVRZNOPCK-UHFFFAOYSA-N
Synonym 4-pyridinemethaneamine, 4-aminomethyl pyridine, 4-pyridinemethanamine, 4-pyridylmethylamine, 4-picolylamine, 4-aminomethylpyridine, 4-picolinylamine, 4-pyridinemethylamine, 1-pyridin-4-ylmethanamine, 4-picolinamineShow MoreShow Less
PubChem CID 77317
IUPAC Name pyridin-4-ylmethanamine
SMILES C1=CN=CC=C1CN
Specifications
Melting Point -8°C
Density 1.069
Boiling Point 230°C
Flash Point 108°C (226°F)
Odor Amine-like
Refractive Index 1.5515
Quantity 25 g
UN Number UN2735
Beilstein 108057
Sensitivity Air Sensitive
Formula Weight 108.14
Percent Purity ≥97%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456853 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog NumberL06345.14,L06345.22
Lot NumberA0411493 Certificate TypeCertificate of Analysis Date27/07/2023 Catalog NumberL06345.14,L06345.22
Lot Number10147022 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL06345.14,L06345.22
Lot Number10183502 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL06345.14,L06345.22
Lot Number5010F23W Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL06345.14,L06345.22
RUO – Research Use Only
Product Content Correction
Product Title
4-(Aminomethyl)pyridine, 97+%, Thermo Scientific Chemicals >