missing translation for 'onlineSavingsMsg'
Learn More
4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 10378600

4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10378600
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10378600

Brand: Thermo Scientific Acros 228050050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2498-50-2
Molecular Formula C7H9N3·2HCl
Molecular Weight (g/mol) 208.09
MDL Number MFCD00013001
InChI Key GHEHNICLPWTXJC-UHFFFAOYSA-N
Synonym 4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
PubChem CID 75626
ChEBI CHEBI:10618
IUPAC Name 4-aminobenzenecarboximidamide;dihydrochloride
SMILES C1=CC(=CC=C1C(=N)N)N.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >300°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NC6H4C(=NH)NH2·2HCl
Quantity 5 g
Formula Weight 208.09
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Aminobenzamidine dihydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458981 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number228050250,228050050
Lot NumberA0459771 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number228050010,228050250,228050050
Lot NumberA0384228 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number228050010,228050250,228050050
Lot NumberA0437348 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog Number228050010,228050250,228050050
Lot NumberK05U011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number228050250,228050050
compliance-icons
Product Identifier
  • 4-Aminobenzamidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.