missing translation for 'onlineSavingsMsg'
Learn More

4',5-Dihydroxyflavone, 95%, Thermo Scientific™

Product Code. 11426620
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6665-67-4
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00017691
InChI Key OKRNDQLCMXUCGG-UHFFFAOYSA-N
Synonym 4',5-dihydroxyflavone, 5,4'-dihydroxyflavone, 5-hydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, 5-hydroxy-2-4-hydroxyphenyl chromen-4-one, 5,4-dihydroxyflavone, 4h-1-benzoicpyran-4-one, 5-hydroxy-2-4-hydroxyphenyl, 4,5-dihydroxyflavone, 4h-1-benzopyran-4-one, 5-hydroxy-2-4-hydroxyphenyl
PubChem CID 165521
IUPAC Name 5-hydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC2=C(C(=C1)O)C(=O)C=C(O2)C3=CC=C(C=C3)O

Specifications

Quantity 1 g
Formula Weight 254.24
Percent Purity 95%
Chemical Name or Material 4',5-Dihydroxyflavone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'