missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1-Methyl-1H-pyrazol-3-yl)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 4-1-methyl-1h-pyrazol-3-yl aniline, 4-1-methyl-1h-pyrazol-3-yl-phenylamine, 4-1-methylpyrazol-3-yl aniline, 1-methyl-3-4-aminophenyl-1h-pyrazole, benzenamine, 4-1-methyl-1h-pyrazol-3-yl, 4-1-methylpyrazol-3-yl phenylamine, 4-1-methyl-1h-pyrazol-3-yl-aniline, 4-1-methyl-1h-pyrazol-3-yl phenylamine, benzenamine,4-1-methyl-1h-pyrazol-3-yl |
Formula Weight | 173.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1-Methyl-1H-pyrazol-3-yl)aniline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(1-Methyl-1H-pyrazol-3-yl)aniline, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction