Learn More
3M™ Versaflo™ M-400 Series PAPR Inner Collar
Click to view available options
:
Inner collar
Description
- The M-444 Inner Collar is a required part of a M-400 system.
- 3M Catalog# M-444
Specifications
Specifications
| Color | Black |
| Disposable | Reusable |
| Product Type | Inner Collar |
| For Use With (Application) | Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding |
| For Use With (Equipment) | M-400 Helmet Shrouds |
| Model | M-444 |