Learn More
3M™ Versaflo™ Peel-Off Visor Cover for Large S-Series Hood and Headcovers
Click to view available options
:
Peel-off visor cover; Medium/Large
Description
- Transparent covering to hood
- Protect from contaminants and scratches
- 3M Catalog# S-920L
Specifications
Specifications
| Color | Clear |
| Disposable | Reusable |
| Product Type | Lens Cover |
| Size | Large |
| For Use With (Application) | Protects Visor From Contaminants and Scratches |
| For Use With (Equipment) | Large Headcovers and Hoods, S133L, S133LB, S333L, S433L, S533L |
| Model | S-920 L |