Learn More
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
- Resilient silicone faceseal offers extended facepiece life even in high-heat environments
- Quick latch design eliminates need to remove a hardhat or faceshield when lowering or raising the mask
- Sleek, low-profile design
- Special valve cover helps to direct exhaled breath and moisture downward
- 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
- Fits securely and comfortably on the head
- Compatible with all 3M™ bayonet-style filters and cartridges:
- 6000 Series Cartridges
- 2000, 2200, 7000 or 5000 Series Filters
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Certifications (OSHA) | OSHA 29 CFR Part 1920.134 |
| Connection Type | Bayonet |
| Color | Gray, Teal |
| Product Type | Half-Mask Respirator |
| Size | Small |
| Facepiece Material | Silicone |
| For Use With (Application) | Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning |
| For Use With (Equipment) | Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters |
| Includes | Quick Latch |
| Show More |