Learn More
3M™ Head Harness Assembly for 7500 Series Respirators
Click to view available options
:
Head harness assembly
Description
The 3M™ Head Harness Assembly 7581, Respiratory Protection System Component, works with 3M Half Facepieces 7501, 7502, and 7503 (all sold separately).
- Approved 3M replacement part
- Designed for use with the 3M Ultimate FX Full Facepiece Reusable Respirators (sold separately)
- Head harness assembly can extend service life of facepiece
- Easy installation allows for quick replacement
- Replaceable part can potentially result in respiratory equipment cost savings
- 3M Catalog# 7581
Specifications
Specifications
| Product Type | Head Harness Assembly |
| For Use With (Application) | Demolition, facility maintenance, grinding, sanding, welding |
| For Use With (Equipment) | Half facepiece respirator 7501, 7502 and 7503 |
| Model | 7581 |
| Dimensions (L x W x H) | 16.4 x 11.4 x 7.3 in. (41.6 x 28.9 x 18.5 cm) |
| Length (English) | 16.4 in. |
| Length (Metric) | 41.6 cm |
| Width (English) | 11.4 in. |
| Width (Metric) | 28.9 cm |
| Height (English) | 7.3 in. |
| Show More |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion