Learn More
Euro Protection™ 3D Flat Fold Masks FFP3 NR D
Click to view available options
:
20/Box, 12 Boxes/Ctn
Description
- Multi-layer design
- Soft inner layer
- Adjustable tightening elastic
- Nose clip
- Nose seal/face seal
Specifications
Specifications
| Certifications/Compliance | EN 149:2001 + A1:2009 |
| Disposable | Single-use |
| Material | SMS Polypropylene |
| Size | One Size |
| Filtration Layer Material | Melt-Blown electrostatic polypropylene |
| Includes | Nose clip |
| Latex Free | Yes |
| Material Headstrap | Latex-free synthetic rubber |
| Material (Liner) | Soft polypropylene |
| Nose Piece Material | Reinforced Polyethylene |
| Show More |