Learn More
Nickel Electro™ Stainless Steel Combustion Spoon
Shop All Nickel-Electro Products
Click to view available options
Length (Metric):
220 mm
Unit Size:
Pack of 6
Description
- Combustion Spoon/Deflagrating Spoon
- 304 spec stainless steel
- Nuffield pattern
- Heavy gauge stainless steel wire
- Designed for use in test tubes stem is shaped to prevent contact with wall of test tube.
- Testing for Acid rain
Specifications
Specifications
| Quantity | 6 Pk. |
| Description | Combustion spoon |
| Autoclavable | Autoclavable |
| Product Type | Spoon |
| For Use With (Application) | For Use in Test Tubes Stem is Shaped to Prevent Contact with Wall of Test Tube |
| Blade Material | Stainless Steel |
| Handle Material | Stainless Steel |
| Material | Stainless Steel |
| Length (Metric) | 220 mm |