missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)cyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 15466997
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15466997

Brand: Thermo Scientific Alfa Aesar H34087.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-36-4
Molecular Formula C7H9F3O
Molecular Weight (g/mol) 166.14
MDL Number MFCD00034662
InChI Key YHADNJOEQWBXAR-UHFFFAOYNA-N
Synonym 3-trifluoromethyl cyclohexanone, 3-trifluoromethyl cyclohexan-1-one, cyclohexanone, 3-trifluoromethyl, 3-trifluoromethyl-cyclohexanone, 3-trifluoromethyl-1-cyclohexanone, 1-oxo-3-trifluoromethyl cyclohexane
PubChem CID 2779355
IUPAC Name 3-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCCC(=O)C1

Specifications

Boiling Point 82°C (29 mmHg)
Quantity 1 g
UN Number UN1224
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)cyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.