missing translation for 'onlineSavingsMsg'
Learn More

3-methylthiophene-2-carbonyl Chloride, 98%, Thermo Scientific™

Product Code. p-3211943 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 10197832

Brand: Thermo Scientific Acros 380400020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 61341-26-2
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00130091
InChI Key AJSVPEVDFBYRCH-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
PubChem CID 2737605
IUPAC Name 3-methylthiophene-2-carbonyl chloride
SMILES CC1=C(SC=C1)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 216°C to 220°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5830 to 1.585
Quantity 2 g
Formula Weight 160.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene-2-carbonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylthiophene-2-carbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.