missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carbonitrile, 96%

Product Code. p-7021108
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11400903

Brand: Thermo Scientific Alfa Aesar A13305.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55406-13-8 | C6H5NS | 123.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55406-13-8
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.173
MDL Number MFCD00052086
InChI Key ALZHYEITUZEZMT-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonitrile, 2-cyano-3-methylthiophene, acmc-1axot, 3-methyl-thiophene-2-carbonitrile, 3-methyl-2-thiophenecarbonitrile #
PubChem CID 521617
IUPAC Name 3-methylthiophene-2-carbonitrile
SMILES CC1=C(SC=C1)C#N

Specifications

Boiling Point 91°C to 93°C (12 mmHg)
Refractive Index 1.553
Quantity 1 g
UN Number UN3276
Beilstein 5495606
Formula Weight 123.18
Percent Purity 96%
Chemical Name or Material 3-Methylthiophene-2-carbonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.