missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Isobutyl-1-methylxanthine, 99+%

€ 157.79 - € 575.87

Chemical Identifiers

CAS 28822-58-4
Molecular Formula C10H14N4O2
Molecular Weight (g/mol) 222.25
MDL Number MFCD00005584
InChI Key APIXJSLKIYYUKG-UHFFFAOYSA-N
Synonym 3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
PubChem CID 3758
ChEBI CHEBI:43253
SMILES CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10617572
View Documents
Thermo Scientific Chemicals
228420010
1 g Glass bottle
€ 157.79
1g
Please to purchase this item. Need a web account? Register with us today!
10257553
View Documents
Thermo Scientific Chemicals
228420050
5 g Glass bottle
€ 575.87
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28822-58-4
222.25
APIXJSLKIYYUKG-UHFFFAOYSA-N
3758
CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O
C10H14N4O2
MFCD00005584
3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
CHEBI:43253

Specifications

28822-58-4
100.0
White to Beige
99+%
MFCD00005584
3-isobutyl-1-methylxanthine, ibmx, isobutylmethylxanthine, 1-methyl-3-isobutylxanthine, methylisobutylxanthine, 3-isobutyl-1-methyl-1h-purine-2,6 3h,7h-dione, xanthine, 3-isobutyl-1-methyl, 3-isobutyl-1-methyxanthine, 1h-purine-2,6-dione, 3,7-dihydro-1-methyl-3-2-methylpropyl, methyl-isobutylxanthine
APIXJSLKIYYUKG-UHFFFAOYSA-N
1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
3758
222.25
Fluffy Crystalline Powder
99.0
200.0°C to 203.0°C
Authentic
C10H14N4O2
26, IV, 2350
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,2-propanol
CC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O
222.25
CHEBI:43253
99+%
3-Isobutyl-1-methylxanthine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 249-259-3

Product Identifier
  • 3-Isobutyl-1-methylxanthine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only