missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminopropylamine, 99%

Product Code. p-3736273 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
2.5 L
250mL
5 L
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10234240

Brand: Thermo Scientific Acros 115902500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-55-7 | C5H14N2 | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-55-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008216
InChI Key IUNMPGNGSSIWFP-UHFFFAOYSA-N
Synonym 3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine
PubChem CID 7993
IUPAC Name N',N'-dimethylpropane-1,3-diamine
SMILES CN(C)CCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 0.8100g/mL
Boiling Point 133.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)3NH2
Refractive Index 1.4340 to 1.4360
Quantity 250 mL
Beilstein 04, III, 554
Fieser 01,274
Specific Gravity 0.81
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Viscosity 1.6 mPa.s (20°C)
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Dimethylaminopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Dimethylaminopropylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.