missing translation for 'onlineSavingsMsg'
Learn More
3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™ Product Code.: 10198312

3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™

Product Code. 10198312
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10198312

Brand: Thermo Scientific Maybridge SEW01730DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 184970-24-9
Molecular Formula C7H5ClN2OS
Molecular Weight (g/mol) 200.64
InChI Key KHJYOJVUQYWASE-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-thiophen-3-yl-1,2,4-oxadiazole, 3-chloromethyl-5-3-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-3-yl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-thienyl, 3-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
PubChem CID 2797043
IUPAC Name 3-(chloromethyl)-5-thiophen-3-yl-1,2,4-oxadiazole
SMILES C1=CSC=C1C2=NC(=NO2)CCl

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 200.65
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-(3-thienyl)-1, 2, 4-oxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.