Learn More
3-Bromo-7-nitroindazole, Tocris Bioscience™
Product Code. 18025872
Click to view available options
Quantity:
10 mg
50 mg
Selective nNOS inhibitor
Potent inhibitor of rat cerebellar nitric oxide synthase; more potent than 7-nitroindazole (Cat. No. 0602).
Chemical Identifiers
CAS 74209-34-0
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.03
MDL Number MFCD00159910
InChI Key NFSTZPMYAZRZPC-UHFFFAOYSA-N
Synonym 3-bromo-7-nitroindazole, 3-bromo-7-nitro-1h-indazole, 1h-indazole,3-bromo-7-nitro, brni, 3-bromo-7ni, 3-br-7-nitroindazole, tocris-0735, lopac-b-2050, d0u6zt, d0x5qj
PubChem CID 1649
IUPAC Name 3-bromo-7-nitro-2H-indazole
SMILES [O-][N+](=O)C1=CC=CC2=C(Br)NN=C12
Specifications
Quantity 50 mg
Formula Weight 242.03
Percent Purity >98%
Chemical Name or Material 3-Bromo-7-nitroindazole