missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-2,2-dimethyl-1-propanol, 95%, Thermo Scientific Chemicals Product Code.: 11308435

3-Amino-2,2-dimethyl-1-propanol, 95%, Thermo Scientific Chemicals

Product Code. 11308435
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11308435

Brand: Thermo Scientific Alfa Aesar L10429.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

Solubility
Soluble in Dichloromethane, Chloroforn, Ether and Methanol

Notes
Hygroscopic,Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26734-09-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00059847
InChI Key FNVOFDGAASRDQY-UHFFFAOYSA-N
Synonym 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
PubChem CID 117326
IUPAC Name 3-amino-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN)CO

Specifications

Melting Point ∼70°C
Quantity 5 g
Beilstein 1731900
Sensitivity Hygroscopic
Solubility Information Soluble in Dichloromethane,Chloroforn,Ether and Methanol
Formula Weight 103.17
Percent Purity 95%
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-2,2-dimethyl-1-propanol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.