missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dihydroxyflavone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
Melting Point | 231°C to 234°C |
Quantity | 5 g |
Solubility Information | Soluble in water. |
Formula Weight | 254.24 |
Percent Purity | 97% |
Chemical Name or Material | 3,6-Dihydroxyflavone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'