Learn More
Thermo Scientific Maybridge 25GR 4-Oxo-4-(4-toluidino)but-2-enoic acid, 97%
Brand: Thermo Scientific Maybridge S01331EB
25GR 4-Oxo-4-(4-toluidino)but-2-enoic acid, 97%
This item is not returnable.
View return policy
Description
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Oxo-4-(4-toluidino)but-2-enoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Product Title
Thermo Scientific Maybridge 25GR 4-Oxo-4-(4-toluidino)but-2-enoic acid, 97% >