Learn More
B Braun™ 250 mL Braunoderm™ Preoperation Skin Disinfectant
A ready-to-use, quick-acting skin disinfectant for use prior to surgery.
Brand:  B Braun™ 3881105
Product Code. 15813692
  • € 260.00 / Pack of 20
Explore more special offers
Description
Description
  • Combined from alcohol and PVP-iodine
  • For skin disinfection in case of surgical procedures or aspiration
  • Short exposure time from 15 seconds
  • Long-term active
  • Easy to spot disinfected skin areas
  • Active against bacteria (including MRSA, TbB) and fungal infestation, as well as viruses such as vaccinia and polio
  • DGHM/VAH listed
Specifications
Specifications
Skin Disinfectant
Alcohol and PVP-iodine solution
20/Pk.
DGHM/VAH-listed
Pre-surgical skin disinfection
250 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers