Learn More
Thermo Scientific™ 24-Place Fixed-Angle Rotors with Covers
Product Code. p-318446 Shop All Thermo Scientific Products
Click to view available options
Max. Speed:
15,000 rpm
Max. RCF:
21,890 x g
Angle:
45°
Unit Size:
Each
Product Code. 10593851
Brand: Thermo Scientific™ 75007593
Specifications
Type Microliter Rotor
Capacity 24 x 1.5/2 mL
Max. RCF 21,890 x g
Max. Speed 15,000 rpm
Angle 45°