Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ASTM Certificate
Product Code. 10476621 Shop All Retsch Products
Click to view available options
Pore Size:
100 mm
106 mm
125 mm
Unit Size:
Each
Product Code. 10476621
Brand: RETSCH 60.150.106000
Only null left
Specifications
Quantity 1Each
Pore Size 106 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 200 Tap
Height (Metric) 50.8mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 203.2 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel