Learn More
Endecotts 200mm sieve, 4.75mm aperture, stainless steel
Brand:  Endecotts 200SIW4.75
200mm sieve, 4.75mm aperture, stainless steel
Product Code. 11547464
  • € 177.86 / Each
Estimated Shipment: 17-07-2025
to see stock.
Explore more special offers
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Endecotts 200mm sieve, 4.75mm aperture, stainless steel >