Learn More
Ismatec™ 2-Stop Tubing, Tygon HC F-4040-A
Product Code. 10636713
Click to view available options
Tubing Size:
I.D. 0.38 mm
I.D. 1.42 mm
I.D. 1.52 mm
Unit Size:
Pack of 12
Product Code. 10636713
Brand: Ismatec™ SC0157
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Special tubing for hydrocarbons, petroleum products, and distillates.
  • Specially formulated to transport hydrocarbons, petroleum products and distillates.
  • Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.
  • Low gas permeability.
Specifications
Color Code Orange-Green
Diameter (Metric) Inner 0.38 mm
Material Tygon HC
Overall Length (Metric) 400 mm
For Use With (Application) Pumps that incorporate cassettes into the pumphead design. Hydrocarbons, petroleum products, and distillates.
Min. Operating Temperature (Metric) −37°C
Packaging Quantity 12/Pk
Resistance Properties Acids: Limited, Alkaline solutions: Not rec., Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Tubing Size I.D. 0.38 mm
Color Amber
Max. Operating Temperature (Metric) 74°C