Learn More
Ismatec™ 2-Stop Tubing, Tygon E-LFL
Click to view available options
Tubing Size:
I.D. 0.38 mm
I.D. 0.64 mm
I.D. 0.76 mm
I.D. 1.14 mm
I.D. 1.25 mm
I.D. 1.37 mm
I.D. 1.60 mm
I.D. 1.85 mm
I.D. 2.20 mm
Unit Size:
Pack of 12
Description
- DEHP free.
- Longest service life of any clear Tygon tubing material.
- Excellent choice where transparency and good chemical resistance is needed.
- Broad chemical resistance.
- Tasteless.
- Extremely low particulate spallation.
- Meets USP Class VI and FDA criteria.
- Non-Aging.
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR 175.300; US Pharmacopoea Class VI |
| Color Code | Blue-Blue |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 1.60 mm |
| Material | Tygon E-LFL |
| Overall Length (Metric) | 400 mm |
| For Use With (Application) | Pumps that incorporate cassettes into the pumphead design. |
| Min. Operating Temperature (Metric) | −50°C |
| Packaging Quantity | 12/Pk |
| Resistance Properties | Acids: Fair, Alkaline solutions: Fair, Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent Sterile media: Limited |
| Show More |