Learn More
2-Hexyne, 98+%, Thermo Scientific Chemicals
€ 52.36 - € 204.02
Chemical Identifiers
CAS | 764-35-2 |
---|---|
Molecular Formula | C6H10 |
Molecular Weight (g/mol) | 82.146 |
MDL Number | MFCD00009470 |
InChI Key | MELUCTCJOARQQG-UHFFFAOYSA-N |
Synonym | 2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3 |
PubChem CID | 33629 |
IUPAC Name | hex-2-yne |
SMILES | CCCC#CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420527
|
Thermo Scientific Alfa Aesar
B22405.09 |
10 g |
€ 52.36
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430527
|
Thermo Scientific Alfa Aesar
B22405.18 |
50 g |
€ 204.02
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
764-35-2 | |
82.146 | |
MELUCTCJOARQQG-UHFFFAOYSA-N | |
33629 | |
CCCC#CC |
C6H10 | |
MFCD00009470 | |
2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3 | |
hex-2-yne |
Specifications
764-35-2 | |
84°C to 86°C | |
Characteristic | |
MFCD00009470 | |
1731542 | |
MELUCTCJOARQQG-UHFFFAOYSA-N | |
hex-2-yne | |
33629 | |
≥98% |
-90°C | |
−10°C (14°F) | |
C6H10 | |
UN3295 | |
2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3 | |
CCCC#CC | |
82.146 | |
82.15 | |
2-Hexyne |
Safety and Handling
GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
- 2-Hexyne
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Methyl propyl acetylene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.