Learn More
2-Hexyn-1-ol, 97%
Product Code. 11470963
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
Product Code. 11470963
Brand: Thermo Scientific Alfa Aesar A13339.14
Request Bulk or Custom Format
CAS: 764-60-3 | C6H10O | 98.15 g/mol
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Avoid temperature and direct sunlight.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 764-60-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00039540
InChI Key BTDWSZJDLLLTMI-UHFFFAOYSA-N
Synonym 2-hexyn-1-ol, pubchem13064, acmc-209p3i, ksc380a2d, 2-hexyn-1-ol,
PubChem CID 69818
IUPAC Name hex-2-yn-1-ol
SMILES CCCC#CCO
Specifications
Density 0.897
Boiling Point 167°C
Flash Point 68°C (154°F)
Linear Formula CH3(CH2)2C≡CCH2OH
Refractive Index 1.454
Quantity 25 g
Beilstein 1736274
Solubility Information It is soluble in water.
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 2-Hexyn-1-ol