missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyryl chloride, 99%

Product Code. p-3757840 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10742073

Brand: Thermo Scientific Acros 409820050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2736-40-5 | C6H11ClO | 134.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2736-40-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.60
MDL Number MFCD00018812
InChI Key SMUKODJVMQOSAB-UHFFFAOYSA-N
Synonym 2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride
PubChem CID 75954
IUPAC Name 2-ethylbutanoyl chloride
SMILES CCC(CC)C(Cl)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9820g/mL
Boiling Point 138°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5)2CHCOCl
Refractive Index 1.4230 to 1.425
Quantity 5 g
Specific Gravity 0.982
Formula Weight 134.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyryl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethylbutyryl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.