missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine-5'-monophosphate, 99%

€ 120.60 - € 468.33

Chemical Identifiers

CAS 1032-65-1
Molecular Formula C9H14N3O7P
Molecular Weight (g/mol) 307.199
MDL Number MFCD00006546
InChI Key NCMVOABPESMRCP-SHYZEUOFSA-N
Synonym dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
PubChem CID 13945
ChEBI CHEBI:15918
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15466799
View Documents
Thermo Scientific Chemicals
J65244.03
1 g
€ 120.60
1g
Please to purchase this item. Need a web account? Register with us today!
15476799
View Documents
Thermo Scientific Chemicals
J65244.06
5 g
€ 468.33
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2'-Deoxycytidine-5'-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine-5′-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

1032-65-1
307.199
NCMVOABPESMRCP-SHYZEUOFSA-N
13945
[(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
C9H14N3O7P
MFCD00006546
dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
CHEBI:15918
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O

Specifications

1032-65-1
C9H14N3O7P
41062
Soluble in water.
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O
307.199
CHEBI:15918
99%
2'-Deoxycytidine-5'-monophosphate
White
MFCD00006546
dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
NCMVOABPESMRCP-SHYZEUOFSA-N
[(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
13945
307.2
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 213-849-9

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only