missing translation for 'onlineSavingsMsg'
Learn More
2-Chloroaniline, 98+%, Thermo Scientific Chemicals Product Code.: 11402418

2-Chloroaniline, 98+%, Thermo Scientific Chemicals

Product Code. 11402418
2500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11402418

Brand: Thermo Scientific Alfa Aesar A15758.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc baseShow MoreShow Less
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -3°C
Density 1.213
Boiling Point 207°C to 209°C
Quantity 2500 g
Odor Characteristic
Refractive Index 1.588
UN Number UN2019
Beilstein 606077
Merck Index 14,2118
Solubility Information Miscible with alcohol,ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).
Formula Weight 127.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloroaniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253818 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA15758.0E,A15758.0C,A15758.36
Lot Number10219916 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog NumberA15758.0E,A15758.0C,A15758.36
Lot Number10212471 Certificate TypeCertificate of Analysis Date21/07/2023 Catalog NumberA15758.0E,A15758.0C,A15758.36
Lot NumberE15N24 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15758.0E,A15758.0C,A15758.36
Lot Number10116375 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15758.0E,A15758.0C,A15758.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloroaniline, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.