missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4,5-dimethoxycinnamic acid, 97%, Thermo Scientific™ Product Code.: 11452888

2-Bromo-4,5-dimethoxycinnamic acid, 97%, Thermo Scientific™

Product Code. 11452888
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11452888

Brand: Thermo Scientific Alfa Aesar A16862.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151539-52-5
Molecular Formula C11H11BrO4
Molecular Weight (g/mol) 287.109
MDL Number MFCD00017614
InChI Key MSJCWIFRXCLULX-ONEGZZNKSA-N
Synonym 2-bromo-4,5-dimethoxycinnamic acid, 6-bromo-3,4-dimethoxycinnamic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, e-3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 3-2-bromo-4,5-dimethoxyphenyl acrylic acid, 2-propenoic acid, 3-2-bromo-4,5-dimethoxyphenyl, e-3-2-bromo-4,5-dimethoxyphenyl prop-2-enoic acid, 2e-3-2-bromo-4,5-dimethoxyphenyl-2-propenoic acid
PubChem CID 735828
IUPAC Name (E)-3-(2-bromo-4,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C(=C1)C=CC(=O)O)Br)OC

Specifications

Melting Point 253°C to 256°C
Quantity 25 g
Beilstein 3142451
Formula Weight 287.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-dimethoxycinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4,5-dimethoxycinnamic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.