missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropionyl Bromide, 98%

Product Code. p-3735754 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10289540

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20769-85-1 | C4H6Br2O | 229.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20769-85-1
Molecular Formula C4H6Br2O
Molecular Weight (g/mol) 229.9
MDL Number MFCD00000122
InChI Key YOCIJWAHRAJQFT-UHFFFAOYSA-N
Synonym 2-bromo-2-methylpropionyl bromide, 2-bromoisobutyryl bromide, propanoyl bromide, 2-bromo-2-methyl, bibb, 2-bromo-2-methylpropionylbromide, unii-627f9n9dya, 2-bromoisobutyrylbromide, 2-bromo-2-methyl-propionyl bromide, alpha-bromoisobutyryl bromide, 2-bromo-2-methyl-propanoyl bromide
PubChem CID 88685
IUPAC Name 2-bromo-2-methylpropanoyl bromide
SMILES CC(C)(C(=O)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.8600g/mL
Boiling Point 162°C to 164°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CBrCOBr
Refractive Index 1.5070 to 1.5090
Quantity 5 g
Specific Gravity 1.86
Solubility Information Solubility in water: reacts
Formula Weight 229.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropionyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2-methylpropionyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.