missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(trifluoromethoxy)phenyl]ethan-1-one, 97%, Thermo Scientific™

Product Code. p-2697916
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10283422

Brand: Thermo Scientific Maybridge TL00407DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103962-10-3
Molecular Formula C9H6BrF3O2
Molecular Weight (g/mol) 283.044
MDL Number MFCD00052333
InChI Key AOAGGWLQIILIIV-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
PubChem CID 2778691
IUPAC Name 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4C(O)CH2Br
Quantity 1 g
Formula Weight 283.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-[4-(trifluoromethoxy)phenyl]ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.