missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole-6-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15454107
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-85-6
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.21
MDL Number MFCD00054180
InChI Key ZEAKWWWXCZMODH-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole-6-carboxylic acid, 2-aminobenzo d thiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, 2-amino, 2-amino-benzothiazole-6-carboxylic acid, 2-amino-6-benzothiazolecarboxylic acid, asischem b67429, zlchem 188, akos bb-7723, salor-int l481106-1ea, ksc487a1lShow MoreShow Less
PubChem CID 66740
IUPAC Name 2-amino-1,3-benzothiazole-6-carboxylic acid
SMILES NC1=NC2=CC=C(C=C2S1)C(O)=O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 194.21
Percent Purity 95%
Chemical Name or Material 2-Aminobenzothiazole-6-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminobenzothiazole-6-carboxylic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.