Learn More
2-Amino-3-methylpyrazine, 95%, Thermo Scientific™
Product Code. 15497758
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19838-08-5
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD08705770
InChI Key VQPHZDDLWFHRHR-UHFFFAOYSA-N
Synonym 2-amino-3-methylpyrazine, 3-methyl-2-pyrazinamine, pyrazinamine, 3-methyl, 2-pyrazinamine, 3-methyl, 3-methyl-2-aminopyrazine, pubchem19617, acmc-1cacy, 2-pyrazinamine,3-methyl, 3-methylpyrazin-2-ylamine, 2-amino-3-methyl-pyrazine
PubChem CID 302284
IUPAC Name 3-methylpyrazin-2-amine
SMILES CC1=NC=CN=C1N
Specifications
Quantity 250 mg
Formula Weight 109.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-3-methylpyrazine