missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-bromothiophene, 99%

Product Code. p-49423 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10567763

Brand: Thermo Scientific Acros 308330050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5370-25-2 | C6H5BrOS | 205.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5370-25-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD00014528
InChI Key IGBZCOWXSCWSHO-UHFFFAOYSA-N
Synonym 2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
PubChem CID 79335
IUPAC Name 1-(5-bromothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Yellow
Boiling Point 103°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 205.07
Percent Purity 99%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2-Acetyl-5-bromothiophene, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.