missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dihydroxyacetophenone, 97%

Product Code. p-112004 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10204480

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 490-78-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-78-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002343
InChI Key WLDWSGZHNBANIO-UHFFFAOYSA-N
Synonym 2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy
PubChem CID 10279
IUPAC Name 1-(2,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 203.0°C to 208.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3COCH3
Quantity 5 g
Beilstein 08, 271
Solubility Information Solubility in water: insoluble
Formula Weight 152.15
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2', 5'-Dihydroxyacetophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2',5'-Dihydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.