Learn More
2-(4-Methylphenyl)thioacetamide, 90+%, Thermo Scientific™
Product Code. 11376769
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97426-53-4
Molecular Formula C9H11NS
Molecular Weight (g/mol) 165.254
MDL Number MFCD00278201
InChI Key NDIXABMZJKQGHJ-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanethioamide, 2-p-tolyl thioacetamide, maybridge1_003524, 2-p-tolyl ethanethioamide, p-tolyl-thioacetic acid amide, 1-amino-2-4-methylphenyl ethane-1-thione
PubChem CID 2806791
IUPAC Name 2-(4-methylphenyl)ethanethioamide
SMILES CC1=CC=C(C=C1)CC(=S)N
Specifications
Melting Point 111°C to 114°C
Quantity 1 g
Formula Weight 165.25
Percent Purity ≥90%
Chemical Name or Material 2-(4-Methylphenyl)thioacetamide