missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylthiophene, 97%

Product Code. p-4510589 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10337721

Brand: Thermo Scientific Acros 342270010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-16-6 | C6H8S | 112.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-16-6
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00130081
InChI Key BZYUMXXOAYSFOW-UHFFFAOYSA-N
Synonym thiophene, 2,3-dimethyl, 2,3-dimethyl-thiophene, 2,3,dimethylthiophene, pubchem7751, 2.3-dimethylthiophene, dimethylthiophene, 2,3-dirnethylthiophene
PubChem CID 34295
IUPAC Name 2,3-dimethylthiophene
SMILES CC1=C(C)C=CS1

Specifications

Color Colorless to Yellow
Flash Point >1°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5170 to 1.5210
Quantity 1 g
Formula Weight 112.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethylthiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3-Dimethylthiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.