missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione monoxime, 98%

Product Code. p-4510165 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10113250

Brand: Thermo Scientific Acros 150371000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-hydroxyiminobutan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 74°C to 76°C
Color White
Boiling Point 185°C to 186°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(=NOH)COCH3
Quantity 100 g
Beilstein 01,772
Solubility Information (5% in water) Clear to slightly hazy
Formula Weight 101.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Butanedione monoxime
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.