Learn More
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%, Thermo Scientific Chemicals
€ 89.04 - € 311.55
Chemical Identifiers
CAS | 540-84-1 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00008943 |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
PubChem CID | 10907 |
ChEBI | CHEBI:62805 |
IUPAC Name | 2,2,4-trimethylpentane |
SMILES | CC(C)CC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348127
|
Thermo Scientific Alfa Aesar
022901.K2 |
1 L |
€ 89.04
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368127
|
Thermo Scientific Alfa Aesar
022901.M1 |
4 x 1 L |
€ 311.55
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
540-84-1 | |
114.232 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
10907 | |
2,2,4-trimethylpentane |
C8H18 | |
MFCD00008943 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CHEBI:62805 | |
CC(C)CC(C)(C)C |
Specifications
540-84-1 | |
Colorless | |
98°C to 99°C | |
410°C (770°F) | |
Nearly odorless | |
1.391 | |
1696876 | |
Miscible with benzene,toluene,xylene,chloroform,ether,carbon disulfide,carbon tetrachloride,acetone,heptane and oils. Immiscible with water. | |
CC(C)CC(C)(C)C | |
114.232 | |
CHEBI:62805 | |
HPLC | |
2, 2, 4-Trimethylpentane |
°C to 107°C | |
0.692 | |
-7°C (19°F) | |
>99.7% | |
C8H18 | |
MFCD00008943 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
2,2,4-trimethylpentane | |
10907 | |
114.23 | |
Liquid |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Hazard Class: 3, Packing Group: II
missing translation for 'einecsNumber' : 208-759-1
missing translation for 'rtecsNumber' : SA3320000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2,2,4-Trimethylpentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : 2,2,4-Trimethylpentane