missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -107°C |
Density | 0.692g/mL at 25°C |
Boiling Point | 99°C |
Quantity | 500 mL |
Explosion Limit | 0.06 |
Flash Point | −12°C (10.4°F) |
Ignition Point | 745°F |
Odor | Characteristic |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'