missing translation for 'onlineSavingsMsg'
Learn More
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%, Thermo Scientific Chemicals Product Code.: 15400247

1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%, Thermo Scientific Chemicals

Product Code. 15400247
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 15400247

Brand: Thermo Scientific Alfa Aesar J62250.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

Selective inhibitor of nitric oxide-sensitive guanylyl cyclase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.158
MDL Number MFCD00792620
InChI Key LZMHWZHOZLVYDL-UHFFFAOYSA-N
Synonym 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odqShow MoreShow Less
PubChem CID 1456
IUPAC Name [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
SMILES C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

Melting Point 160°C to 170°C
Color White
Quantity 50 mg
Formula Weight 187.15
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX04I043 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberJ62250.MF,J62250.MA
Lot NumberQ22I041 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog NumberJ62250.MF,J62250.MA
Lot NumberY02C035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62250.MF,J62250.MA
Lot NumberL20Z028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62250.MF,J62250.MA
Lot NumberU20H016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62250.MF,J62250.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.