Learn More
Bochem™ 18/10 Stainless Steel Spoons
Click to view available options
Length (Metric):
120 mm
135 mm
180 mm
195 mm
280 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |
| Product Type | Spoon |
| Handle Material | Stainless Steel (18/10) |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 195 mm |