Learn More
Bochem™ 18/10 Stainless Steel Forceps, Blunt Tip
missing translation for 'orderingAttributeHoverText'
Length (Metric):
105 mm
130 mm
145 mm
160 mm
200 mm
250 mm
300 mm
missing translation for 'unitSize'
Each
Pack of 10
Description
- Stainless Steel
- Various lengths
Spécification
Spécification
| Tip Style | Blunt |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 200 mm |
| Product Type | Forceps |
| Quantity (No. per Case) | 1 Pack |
| Type | Forceps |