Learn More
Bochem™ 18/10 Stainless Steel Forceps, Blunt Tip
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Forceps serve a wide variety of functions in the lab.
Brand: Bochem™ 1004
Description
- Stainless Steel
- Various lengths
Specifications
Blunt | |
160 mm | |
1 Pack |
Stainless Steel (18/10) | |
Forceps | |
Forceps |
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Forceps, Blunt Tip > Tip Style: Blunt; Length: 160mm; Quantity: 1/Pk