Learn More
Bochem™ 18/10 Stainless Steel Double Spatulas, Straight
Click to view available options
Length (Metric):
130 mm
150 mm
185 mm
210 mm
250 mm
300 mm
Unit Size:
Each
Description
- Double-ended.
- Round end and flat end.
- 18/10 stainless steel construction.
Specifications
Specifications
| Product Type | Double Ended Spatula |
| Length (Metric) Blade | 50 mm |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 185 mm |