Learn More
Bochem™ 18/10 Stainless Steel Double Spatulas, Straight
Product Code. 10232061
Click to view available options
Length (Metric):
130 mm
150 mm
185 mm
210 mm
250 mm
300 mm
Unit Size:
Each
Product Code. 10232061
Brand: Bochem™ 3104
Only null left
Add to basket
Add to basket
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Double Spatulas serve many uses in the lab.
  • Double-ended.
  • Round end and flat end.
  • 18/10 stainless steel construction.
Specifications
Product Type Double Ended Spatula
Length (Metric) Blade 70 mm
Material Stainless Steel (18/10)
Length (Metric) 250 mm